Skip to Content
Merck
All Photos(1)

Key Documents

JKP6004R

Sigma-Aldrich

6arm-PEG17K

dipentaerythritol core, average Mn 17,000

Synonym(s):

6arm-PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
R(O(CH2CH2O)nH)6
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 17,000

storage temp.

−20°C

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service