Skip to Content
Merck
All Photos(1)

Key Documents

JKA10019

Sigma-Aldrich

8arm-PEG20K

average MN 20,000, Maleimide, tripentaerythritol core, thiol reactive

Synonym(s):

8arm-PEG-Maleimide, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€540.00

€540.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€540.00

About This Item

Linear Formula:
R(O(CH2CH2O)nCH2CH2NHCOCH2CH2C4H2O2N)8
UNSPSC Code:
12162002
NACRES:
NA.23

€540.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

8arm-PEG20K-Maleimide, tripentaerythritol core, average Mn 20,000

form

solid

mol wt

average Mn 20,000

reaction suitability

reactivity: thiol reactive

polymer architecture

shape: 8-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

8arm PEG Maleimide. Crosslinks into hydrogels. MAL is selective for thiol groups on cystein side chains; reacts at pH 5.0-6.5. 8ARM(TP)-PEG with tripentaerythritol core has a lower polydispersity and higher molecular weight accuracy compared with the generic 8ARM-PEG with a hexaglycerin core.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service