Skip to Content
Merck
All Photos(1)

Documents

HTOA200

Oleylamine

≥89%

Synonym(s):

Oleylamine, cis-1-Amino-9-octadecene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2NH2
CAS Number:
Molecular Weight:
267.49
Beilstein:
1723960
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

8 mmHg ( 135 °C)

Assay

≥89%

refractive index

n20/D 1.4596 (lit.)

bp

348-350 °C (lit.)

mp

18-26 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCN

InChI

1S/C18H37N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10H,2-8,11-19H2,1H3/b10-9-

InChI key

QGLWBTPVKHMVHM-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service