Skip to Content
Merck
All Photos(1)

Key Documents

H17856

Sigma-Aldrich

trans-β-Hydromuconic acid

98%

Synonym(s):

trans-3-Hexenedioic acid, 2-Butene-1,4-dicarboxylic acid, Dihydromuconic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€185.00
50 G
€693.00

€185.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€185.00
50 G
€693.00

About This Item

Linear Formula:
HO2CCH2CH=CHCH2CO2H
CAS Number:
Molecular Weight:
144.13
Beilstein:
1723581
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€185.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

195-196 °C (lit.)

SMILES string

[H]\C(CC(O)=O)=C(\[H])CC(O)=O

InChI

1S/C6H8O4/c7-5(8)3-1-2-4-6(9)10/h1-2H,3-4H2,(H,7,8)(H,9,10)/b2-1+

InChI key

YHGNXQAFNHCBTK-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service