This product has a lower working temperature of 20 °C and can tolerate an upper working temperature range of 115-170 °C.
GF81827781
Polyethylene Terephthalate
(polyester, PET, PETP), sheet, thickness 1 mm, size 150 × 150 mm, condition amorphous
Synonym(s):
ES303010
Select a Size
Select a Size
About This Item
Recommended Products
form
sheet
manufacturer/tradename
Goodfellow 818-277-81
size
150 × 150 mm
thickness
1 mm
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
I would like to ask how well the PET sheet can tolerate heat. Thank you!
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service