Skip to Content
Merck
All Photos(2)

Key Documents

GF79962137

Cobalt

foil, 15mm disks, thickness 0.125mm, annealed, 99.9%

Synonym(s):

Cobalt, CO000241

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 799-621-37

resistivity

6.24 μΩ-cm, 20°C

diam. × thickness

15 mm × 0.125 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kian Keat Lee et al.
Biosensors & bioelectronics, 39(1), 255-260 (2012-08-18)
Cobalt oxyhydroxide, CoOOH, nanosheets were prepared via a surface alkaline treatment of cobalt foil at room temperature without using templates and catalysts. The morphology, chemical composition and structures of the nanosheets were characterized by XRD, FTIR and Raman spectroscopy, FESEM
M D Wirt et al.
Biochemistry, 34(15), 5269-5273 (1995-04-18)
We have completed the first direct structural characterization of an enzyme-bound four-coordinate Co(I) intermediate, in this case for the corrinoid/iron-sulfur protein (C/Fe-SP) from Clostridium thermoaceticum. Extended X-ray absorption fine structure and X-ray edge spectroscopy of the active Co(I) state of
Dennis J Paustenbach et al.
Clinical toxicology (Philadelphia, Pa.), 52(2), 98-112 (2013-11-22)
INTRODUCTION. There has been some recent concern regarding possible systemic health effects resulting from elevated blood cobalt concentrations in patients with cobalt containing hip implants. To date there are no blood cobalt criteria to help guide physicians when evaluating an
S Valanarasu et al.
Journal of nanoscience and nanotechnology, 14(6), 4286-4291 (2014-04-18)
Cobalt Oxide (Co3o4) thin films have been successfully coated onto glass substrates at various solution pH by sol-gel spin coating technique. The film thickness was estimated using weight gain method and it revealed that the film thickness increased with solution
Lorale J Lalgee et al.
Biochimica et biophysica acta, 1838(5), 1247-1254 (2014-02-11)
Three amphiphilic cobalt-cage congeners bearing a diaza-crown bridge and varying alkyl chains (1:2:3; n = 12, 16, 18) have been assessed for their ion transport across planar lipid bilayer membranes. In symmetrical electrolyte solutions, a range of ion transport activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service