Skip to Content
Merck
All Photos(2)

Key Documents

GF78384437

Magnesium

wire reel, 2m, diameter 0.25mm, as drawn, 99.9+%

Synonym(s):

Magnesium, MG005115

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
MDL number:
UNSPSC Code:
12141502
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 621 °C)

Assay

99.9%

form

wire

autoignition temp.

950 °F

manufacturer/tradename

Goodfellow 783-844-37

resistivity

4.46 μΩ-cm, 20°C

L × diam.

2 m × 0.25 mm

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Castiglioni et al.
Nutrients, 5(8), 3022-3033 (2013-08-06)
A tight control of magnesium homeostasis seems to be crucial for bone health. On the basis of experimental and epidemiological studies, both low and high magnesium have harmful effects on the bones. Magnesium deficiency contributes to osteoporosis directly by acting
Feng-Yen Li et al.
Blood, 123(14), 2148-2152 (2014-02-20)
Epstein-Barr virus (EBV) is an oncogenic gammaherpesvirus that infects and persists in 95% of adults worldwide and has the potential to cause fatal disease, especially lymphoma, in immunocompromised hosts. Primary immunodeficiencies (PIDs) that predispose to EBV-associated malignancies have provided novel
Stella Lucia Volpe
Advances in nutrition (Bethesda, Md.), 4(3), 378S-383S (2013-05-16)
Magnesium is the fourth most abundant mineral and the second most abundant intracellular divalent cation and has been recognized as a cofactor for >300 metabolic reactions in the body. Some of the processes in which magnesium is a cofactor include
Carlos M Campos et al.
International journal of molecular sciences, 14(12), 24492-24500 (2013-12-20)
The introduction of metallic drug-eluting stents has reduced the risk of restenosis and widened the indications of percutaneous coronary intervention in treatment of coronary artery disease. However, this medical device can induce hypersensitive reaction that interferes with the endothelialization and
Parisa Asghari et al.
Circulation research, 115(2), 252-262 (2014-05-03)
Single-tilt tomograms of the dyads in rat ventricular myocytes indicated that type 2 ryanodine receptors (RYR2s) were not positioned in a well-ordered array. Furthermore, the orientation and packing strategy of purified type 1 ryanodine receptors in lipid bilayers is determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service