GF77502304
Phosphor bronze
foil, thickness 0.08 mm, Cu94%/Sn 6%, length 1 m, coil width 0.4 mm
Synonym(s):
CU050247
About This Item
form
foil
manufacturer/tradename
Goodfellow 775-023-04
L
1 m
coil width
0.4 mm
thickness
0.08 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service