GF68352330
Molybdenum
foil, 6mm disks, thickness 0.75mm, annealed, 99.9%
Synonym(s):
Molybdenum, MO000385
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99.9%
form
foil
manufacturer/tradename
Goodfellow 683-523-30
resistivity
5.0 μΩ-cm, 20°C
diam. × thickness
6 mm × 0.75 mm
bp
4612 °C (lit.)
mp
2617 °C (lit.)
density
10.3 g/mL at 25 °C (lit.)
SMILES string
[Mo]
InChI
1S/Mo
InChI key
ZOKXTWBITQBERF-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dalton transactions (Cambridge, England : 2003), (9)(9), 1298-1303 (2004-07-15)
New molybdenum(VI) nitride oxides were synthesised by the reaction of strontium nitride and calcium nitride with molybdenum foil at high temperature in sealed stainless steel crucibles. The reactions yielded single crystalline products determined by X-ray diffraction to form complex structures
Nanotechnology, 19(26), 265605-265605 (2008-07-02)
Nanocrystalline lanthanum hexaboride (LaB(6)) films have been deposited on molybdenum foil by the pulsed laser deposition (PLD) technique. The as-deposited films were characterized by x-ray diffraction (XRD), scanning electron microscopy (SEM) and atomic force microscopy (AFM). The XRD pattern shows
The Review of scientific instruments, 79(10), 10F125-10F125 (2008-12-03)
Two modular arrays of Langmuir probes designed to handle a heat flux of up to 25 MW/m(2) for 10 s exposures have been installed in the lower divertor target plates of the DIII-D tokamak. The 20 pyrolytic graphite probe tips
The Science of the total environment, 435-436, 96-106 (2012-08-01)
In a regulatory context, bioaccumulation or bioconcentration factors are used for considering secondary poisoning potential and assessing risks to human health via the food chain. In this paper, literature data on the bioaccumulation of molybdenum in the aquatic organisms are
Metal ions in life sciences, 13, 415-450 (2014-01-29)
Molybdenum is an essential trace element and crucial for the survival of animals. Four mammalian Mo-dependent enzymes are known, all of them harboring a pterin-based molybdenum cofactor (Moco) in their active site. In these enzymes, molybdenum catalyzes oxygen transfer reactions
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service