Skip to Content
Merck
All Photos(2)

Key Documents

GF65783784

Lead

wire reel, 2m, diameter 0.25mm, as drawn, 99.95%

Synonym(s):

Lead, PB005111

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€301.00

€301.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€301.00

About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

€301.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99.95%

form

wire

manufacturer/tradename

Goodfellow 657-837-84

resistivity

20.65 μΩ-cm

L × diam.

2 m × 0.25 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reply: Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Karsten Witt et al.
Brain : a journal of neurology, 137(Pt 5), e275-e275 (2014-02-20)
Min-Yi Huang et al.
Ecotoxicology and environmental safety, 104, 310-316 (2014-04-15)
Lead (Pb) is a common heavy metal in the natural environment, but its concentration has been increasing alongside widespread industrial and agricultural development in China. The dark-spotted frog Pelophylax (formerly Rana) nigromaculata (Anura: Ranidae) is distributed across East Asia and
Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Takashi Morishita et al.
Brain : a journal of neurology, 137(Pt 5), e274-e274 (2014-02-13)
Chang-Sun Sim et al.
Environmental research, 130, 1-6 (2014-02-08)
We present data from the Korean National Health and Nutritional Examination Survey (KNHANES) 2010-11 on the distribution of blood lead levels, and examine their association with iron deficiency in a representative sample of the adolescent Korean population. This study was
Shaoxin Huang et al.
Toxicology letters, 226(3), 277-284 (2014-03-07)
Since lead accumulation is toxic to cells, its excretion is crucial for organisms to survive the toxicity. In this study, mouse testis sertoli (TM4) and Mrp1 lower-expression TM4-sh cells were used to explore the lead accumulation characteristics, and the role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service