Skip to Content
Merck
All Photos(1)

Key Documents

GF57602673

Copper/Beryllium

rod, Cu 98%/Be 2%, 20 mm diameter, length 50 mm, temper hard

Synonym(s):

CU077950

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
31152304
NACRES:
NA.23
Pricing and availability is not currently available.

form

rod

manufacturer/tradename

Goodfellow 576-026-73

L

50 mm

diameter

20 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF96573842GF85570803GF05952933
form

rod

form

rod

form

rod

form

rod

L

50 mm

L

500 mm

L

100 mm

L

200 mm

diameter

20 mm

diameter

20 mm

diameter

25 mm

diameter

25 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service