GF56940161
Magnesium
rod, 12.7 mm diameter, length 500 mm, purity 99.9%
Synonym(s):
MG007924
About This Item
vapor pressure
1 mmHg ( 621 °C)
Assay
99.9%
form
rod
autoignition temp.
950 °F
manufacturer/tradename
Goodfellow 569-401-61
resistivity
4.46 μΩ-cm, 20°C
L
500 mm
diameter
12.7 mm
bp
1090 °C (lit.)
mp
648 °C (lit.)
density
1.74 g/mL at 25 °C (lit.)
SMILES string
[Mg]
InChI
1S/Mg
InChI key
FYYHWMGAXLPEAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service