Skip to Content
Merck
All Photos(1)

Key Documents

GF56924466

Bismuth-lead alloy, Bi55.5Pb44.5

100g lump, 100mm max. lump size

Synonym(s):

BI076100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141706
NACRES:
NA.23
Product GF56924466 is not currently sold in your country. Contact Technical Service

form

lumps

manufacturer/tradename

Goodfellow 569-244-66

weight

100 g

max. lump size

100 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Lact. - Repr. 1A - STOT RE 1 Oral - STOT RE 2

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service