Skip to Content
Merck
All Photos(1)

Key Documents

GF50839218

Titanium/Aluminum/Vanadium

rod, Ti 90%/Al 6%/V 4%, 2.0 mm diameter, length 200 mm, temper annealed

Synonym(s):

TI017905

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€182.00
5 EA
€199.00
1 EA
€246.00

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 EA
€182.00
5 EA
€199.00
1 EA
€246.00

About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

form

rod

manufacturer/tradename

Goodfellow 508-392-18

L

200 mm

diameter

2.0 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service