GF44282268
Niobium
foil, thickness 0.025 mm, 4 mm diameter, purity 99.9%
Synonym(s):
NB000301, Columbium
About This Item
Assay
99.9%
form
foil
manufacturer/tradename
Goodfellow 442-822-68
resistivity
13-16 μΩ-cm, 20°C
diam.
4 mm
thickness
0.025 mm
diameter
4 mm
bp
4742 °C (lit.)
mp
2468 °C (lit.)
density
8.57 g/mL at 25 °C (lit.)
SMILES string
[Nb]
InChI
1S/Nb
InChI key
GUCVJGMIXFAOAE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service