GF36625978
Polyphenylenesulfide - 40% Glass Fiber Reinforced
(PPS - 40% GFR), granule, 3 mm nominal granule size, weight 200 g
Synonym(s):
SU316300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
granular
manufacturer/tradename
Goodfellow 366-259-78
weight
200 g
nominal granule size
3 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service