Skip to Content
Merck
All Photos(2)

Documents

GF33636753

Molybdenum

tube, 200mm, outside diameter 3.15mm, inside diameter 2.39mm, wall thickness 0.38mm, 99.9%

Synonym(s):

Molybdenum, MO007350

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

Assay

99.90%

form

tubes

manufacturer/tradename

Goodfellow 336-367-53

resistivity

5.0 μΩ-cm, 20°C

L × wall thickness

200 mm × 0.38 mm

O.D. × I.D.

3.15 mm × 2.39 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service