GF33164614
Hafnium
foil, 4mm disks, thickness 0.0125mm, 97%
Synonym(s):
Hafnium, HF000170
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
MDL number:
UNSPSC Code:
12141718
PubChem Substance ID:
NACRES:
NA.23
Assay
97%
form
foil
manufacturer/tradename
Goodfellow 331-646-14
resistivity
29.6 μΩ-cm, 0°C
diam. × thickness
4 mm × 0.0125 mm
bp
4602 °C (lit.)
mp
2227 °C (lit.)
density
13.3 g/cm3 (lit.)
SMILES string
[Hf]
InChI
1S/Hf
InChI key
VBJZVLUMGGDVMO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service