Skip to Content
Merck
All Photos(2)

Key Documents

GF30163148

Iron

wire reel, 10m, diameter 0.5mm, as drawn, 99.95%

Synonym(s):

Iron, FE005153

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€735.00

€735.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€735.00

About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

€735.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99.95%

form

wire

manufacturer/tradename

Goodfellow 301-631-48

resistivity

9.71 μΩ-cm

L × diam.

10 m × 0.5 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF48022152GF05454132GF17319191
Iron wire reel, 10m, diameter 0.5mm, as drawn, 99.95%

GF30163148

Iron

Iron wire reel, 2m, diameter 0.5mm, hard, 99.99+%

GF48022152

Iron

Iron wire reel, 1m, diameter 0.5mm, as drawn, 99.95%

GF05454132

Iron

Iron wire reel, 1m, diameter 0.25mm, annealed, 99.99+%

GF17319191

Iron

form

wire

form

wire

form

wire

form

wire

manufacturer/tradename

Goodfellow 301-631-48

manufacturer/tradename

Goodfellow 480-221-52

manufacturer/tradename

Goodfellow 054-541-32

manufacturer/tradename

Goodfellow 173-191-91

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

50 years ago in the Journal of Pediatrics: Iron metabolism in premature infants: II. Prevention of iron deficiency.
Pamela J Kling
The Journal of pediatrics, 164(4), 768-768 (2014-03-22)
Laura M van Staalduinen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), 5171-5176 (2014-04-08)
The enzymes PhnY and PhnZ comprise an oxidative catabolic pathway that enables marine bacteria to use 2-aminoethylphosphonic acid as a source of inorganic phosphate. PhnZ is notable for catalyzing the oxidative cleavage of a carbon-phosphorus bond using Fe(II) and dioxygen
Thomas A Russo et al.
Infection and immunity, 82(6), 2356-2367 (2014-03-26)
Hypervirulent (hypermucoviscous) Klebsiella pneumoniae (hvKP) strains are an emerging variant of "classical" K. pneumoniae (cKP) that cause organ and life-threatening infection in healthy individuals. An understanding of hvKP-specific virulence mechanisms that enabled evolution from cKP is limited. Observations by our
Eunna Choi et al.
Infection and immunity, 82(6), 2203-2210 (2014-03-20)
In order to survive inside macrophages, Salmonella produces a series of proteins encoded by genes within Salmonella pathogenicity island 2 (SPI-2). In the present study, we report that Fur, a central regulator of iron utilization, negatively controls the expression of
Hypoferremia of infection: a double-edged sword?
Kristen L Lokken et al.
Nature medicine, 20(4), 335-337 (2014-04-09)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service