GF01547164
Manganin® - resistance alloy, Cu86Mn12Ni2
foil, 4mm disks, 0.5mm thickness, annealed
Synonym(s):
CU060500
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
annealed
foil (disks)
manufacturer/tradename
Goodfellow 015-471-64
size × thickness
4 mm × 0.5 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Manganin is a registered trademark of Isabellenhütte Heusler GmbH & Co. KG.
Manganin is a registered trademark of Isabellenhütte Heusler GmbH & Co. KG.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service