GF00679361
Styrene Butadiene Styrene Block Copolymer
(SBS), granule, 3 mm nominal granule size, weight 1 kg, natural
Synonym(s):
SB306300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352100
Recommended Products
form
granular
manufacturer/tradename
Goodfellow 006-793-61
weight
1 kg
color
natural
nominal granule size
3 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service