Skip to Content
Merck
All Photos(1)

Key Documents

F6312

Sigma-Aldrich

Fmoc-(S)-2-(4-pentenyl)Ala-OH

≥97%, for peptide synthesis

Synonym(s):

(S)-N-Fmoc-α-4-n-pentenylalanine, Fmoc-(S)-2-(4-pentenyl)alanine, Fmoc-(S)-2-amino-2-methyl-hept-6-enoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€273.00
5 G
€988.00

€273.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€273.00
5 G
€988.00

About This Item

Empirical Formula (Hill Notation):
C23H25NO4
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

€273.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Fmoc-(S)-2-(4-pentenyl)Ala-OH, ≥97%

Assay

≥97%

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

<30 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

N([C@](CCCC=C)(C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H25NO4/c1-3-4-9-14-23(2,21(25)26)24-22(27)28-15-20-18-12-7-5-10-16(18)17-11-6-8-13-19(17)20/h3,5-8,10-13,20H,1,4,9,14-15H2,2H3,(H,24,27)(H,25,26)/t23-/m0/s1

InChI key

MRJFPZWLOJOINV-QHCPKHFHSA-N

Application

Fmoc-(S)-2-(4-pentenyl)Ala-OH is a Fmoc-protected amino acid that can be used to create peptide libraries.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z-selective olefin metathesis on peptides: Investigation of side chain influence, preorganization, and guidelines in substrate selection.
O?Leary D J and Grubbs R H
Amino Acids, 5, S13-S13 null
Young-Woo Kim et al.
Nature protocols, 6(6), 761-771 (2011-06-04)
This protocol provides a detailed procedure for the preparation of stapled α-helical peptides, which have proven their potential as useful molecular probes and as next-generation therapeutics. Two crucial features of this protocol are (i) the construction of peptide substrates containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service