Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW448VT

Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting

wide-neck, low-density polyethylene bottle, orange polypropylene closure, capacity 1000 mL

Synonym(s):

WGW448VT

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
€43.20

€43.20


Please contact Customer Service for Availability


Select a Size

Change View
5 EA
€43.20

About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

€43.20


Please contact Customer Service for Availability

material

low-density polyethylene bottle
orange polypropylene closure
wide-neck

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW448VT

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Precision jet tip for fine control

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service