Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW444VT

Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting

wide-neck, low-density polyethylene bottle, yellow polypropylene closure, capacity 1000 mL

Synonym(s):

WGW444VT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

material

low-density polyethylene bottle
wide-neck
yellow polypropylene closure

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW444VT

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Patented Driplok vapour venting valve to prevent solvent drips

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service