Skip to Content
Merck
All Photos(1)

Key Documents

DWKBDH118T

Azlon Acrylic Burette

acrylic, PMP stopcock, PE stopcock, class B, capacity 50 mL

Synonym(s):

BDH118T

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€59.90

€59.90


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€59.90

About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

€59.90


Please contact Customer Service for Availability

material

PE stopcock
PMP stopcock
acrylic

feature

class B

packaging

pkg of 1 ea

manufacturer/tradename

Azlon

storage condition

room temperature

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

AZLON BURETTE ACRYLIC, WITH PMP/PE STOPCOCK, CLASS B, 50 ML

Application

Ideal for educational labs where breakages often occur

Features and Benefits

Robust construction for strength and transparency

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service