Skip to Content
Merck
All Photos(2)

Key Documents

DWK231853606

Reagent Bottle, Wide Neck

wide-neck

Synonym(s):

231853606

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
€240.00

€240.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EACH
€240.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

€240.00


Please contact Customer Service for Availability

material

clear soda-lime glass bottle
wide-neck

Joint

Joints ST/NS neck WITH STOPPER (NS 34/24)

packaging

pkg of 10 ea

manufacturer/tradename

231853606

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Media preparation and cultivation

Features and Benefits

A maximum usage temperature of + 100 °C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3., If the bottle and the stopper have a temperature difference greater than 30 °C, the glass stoppers can become stuck, Easy access to the bottle contents due to the wide diameter of the bottle neck ;

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service