DWK211946854
Duran® Filtering Flasks
Bottle shape, WITH KECK ASSEMBLY SET
Synonym(s):
211946854
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 211946854
capacity
3000 mL
Looking for similar products? Visit Product Comparison Guide
Application
Separations by vacuum filtration, Filtration at reduced pressure, Filtrieren, Vacuum filtration, Safe working under vacuum, Boro 3.3, PBT, PP, VMQ, EPDM
Features and Benefits
Vacuum resistant due to the heavy wall thickness; These filtering flasks fulfil the regulations of the equipment and product safety regulations; Fired-on, highly durable white print; Fired-on, highly durable white print;
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service