Skip to Content
Merck
All Photos(1)

Documents

D97002

Sigma-Aldrich

Diethyl 3-hydroxyglutarate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH(CH2COOC2H5)2
CAS Number:
Molecular Weight:
204.22
Beilstein:
1709448
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

156-157 °C/23 mmHg (lit.)

density

1.103 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(O)CC(=O)OCC

InChI

1S/C9H16O5/c1-3-13-8(11)5-7(10)6-9(12)14-4-2/h7,10H,3-6H2,1-2H3

InChI key

OLLQYIBTJXUEEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tjeerd Pols et al.
Nature communications, 10(1), 4239-4239 (2019-09-20)
One of the grand challenges in chemistry is the construction of functional out-of-equilibrium networks, which are typical of living cells. Building such a system from molecular components requires control over the formation and degradation of the interacting chemicals and homeostasis
Hendrik R Sikkema et al.
Science advances, 6(47) (2020-11-20)
(Micro)organisms are exposed to fluctuating environmental conditions, and adaptation to stress is essential for survival. Increased osmolality (hypertonicity) causes outflow of water and loss of turgor and is dangerous if the cell is not capable of rapidly restoring its volume.
Tjeerd Pols et al.
The FEBS journal, 288(1), 293-309 (2020-04-20)
In cells, the breakdown of arginine to ornithine and ammonium ion plus carbon dioxide is coupled to the generation of metabolic energy in the form of ATP. The arginine breakdown pathway is minimally composed of arginine deiminase, ornithine transcarbamoylase, carbamate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service