Skip to Content
Merck
All Photos(2)

Key Documents

D6388

Sigma-Aldrich

Dimethyl(2-hydroxy-5-nitrobenzyl)sulfonium bromide

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12BrNO3S
CAS Number:
Molecular Weight:
294.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

storage temp.

2-8°C

SMILES string

[Br-].C[S+](C)Cc1cc(ccc1O)[N+]([O-])=O

InChI

1S/C9H11NO3S.BrH/c1-14(2)6-7-5-8(10(12)13)3-4-9(7)11;/h3-5H,6H2,1-2H3;1H

InChI key

VEGVMTIJKACZGO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E McLean et al.
Biochemistry, 40(7), 2194-2200 (2001-05-01)
Brequinar and the active metabolite of leflunomide, A77 1726, have been clearly shown to inhibit human dihydroorotate dehydrogenase (DHODH), but conflicting mechanisms for their inhibition have been reported. DHODH catalyses the conversion of dihydroorotate (DHO) to orotate concurrent with the
Y M Peyser et al.
FEBS letters, 259(2), 346-348 (1990-01-01)
Rabbit skeletal muscle myosin subfragment-1 (S-1) was reacted with dimethyl(2-hydroxy-5-nitrobenzyl)sulfonium bromide (DHNBS) resulting in modification of 0.8 tryptophan residues per S-1. In order to assign the most reactive tryptophan of the 5 S-1 tryptophans, antibodies were raised in rabbits against
G I Karp et al.
Archives of biochemistry and biophysics, 233(2), 712-720 (1984-09-01)
A single tryptophan residue on antithrombin has been modified with dimethyl-(2-hydroxy-5-nitrobenzyl)sulfonium bromide. This alteration led to a 500-fold reduction in the heparin-dependent acceleration of thrombin-modified antithrombin interactions, as well as a 10-fold decrease in the avidity of the modified protease
Structure-function relationship in the allosteric L-lactate dehydrogenases from Lactobacillus casei and Lactobacillus curvatus.
U Mayr et al.
European journal of biochemistry, 126(3), 549-558 (1982-09-01)
Inactivation of the nucleoside transporter of the human erythrocyte by dimethyl (2-hydroxy-5-nitrobenzyl) sulphonium bromide.
S D Thorne et al.
Biochemical Society transactions, 19(4), 418S-418S (1991-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service