Skip to Content
Merck
All Photos(1)

Key Documents

D4202

Sigma-Aldrich

16-Dehydroprogesterone

97%

Synonym(s):

4,16-Pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

185-187 °C (lit.)

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)C(=CC[C@@]24[H])C(C)=O

InChI

1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h6,12,16,18-19H,4-5,7-11H2,1-3H3/t16-,18-,19-,20-,21+/m0/s1

InChI key

VRRHHTISESGZFN-RKFFNLMFSA-N

Gene Information

human ... CYP17A1(1586)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Glass et al.
Journal of steroid biochemistry, 21(1), 65-72 (1984-07-01)
Eubacterium sp. 144 biotransformed 16-dehydroprogesterone by initially hydrating approx 50% to 16 alpha-hydroxyprogesterone. The detection of this reaction was dependent, in part, on the solubility state of 16-dehydroprogesterone and was less extensive when the concentration of methanol was insufficient to
Comparison of the cardiotonic response of ouabain, androstan-3-one, 16-dehydroprogesterone, and conjugated estrogens.
A Woods et al.
Proceedings of the Western Pharmacology Society, 34, 113-115 (1991-01-01)
T L Glass et al.
Applied and environmental microbiology, 49(5), 1146-1153 (1985-05-01)
Suspensions of Eubacterium sp. strain 144, prepared from cells grown with 16-dehydroprogesterone, catalyzed the reduction of this steroid to 17-isoprogesterone at a very low rate. Modifications of the assay to optimize the pH (5.5) and increase the steroid solubility (10%
A E Schindler et al.
Biology of the neonate, 27(3-4), 192-207 (1975-01-01)
Four different methods of isolation and purification were utilized to study steroids in urine of male newborns which was collected during the first 5 days of life. These methods included celite column, ion exchange column and thin-layer chromatography, solvolysis and
Ismael Samudio et al.
Molecular cancer therapeutics, 4(12), 1982-1992 (2005-12-24)
In this study, the antileukemic effects of three isomeric pregnadienedione steroids [i.e., cis-guggulsterone, trans-guggulsterone, and 16-dehydroprogesterone] were investigated in HL60 and U937 cells as well as in primary leukemic blasts in culture. Our results show that all three compounds inhibited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service