Skip to Content
Merck
All Photos(1)

Key Documents

D27608

Sigma-Aldrich

4,4′-Methylenediantipyrine

97%

Synonym(s):

Diantipyrylmethane, Trichachnine monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€37.30

€37.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€37.30

About This Item

Empirical Formula (Hill Notation):
C23H24N4O2 ·H2O
CAS Number:
Molecular Weight:
406.48
Beilstein:
59702
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€37.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

156 °C (dec.) (lit.)

SMILES string

O.CN1N(C(=O)C(CC2=C(C)N(C)N(C2=O)c3ccccc3)=C1C)c4ccccc4

InChI

1S/C23H24N4O2.H2O/c1-16-20(22(28)26(24(16)3)18-11-7-5-8-12-18)15-21-17(2)25(4)27(23(21)29)19-13-9-6-10-14-19;/h5-14H,15H2,1-4H3;1H2

InChI key

OSBQFBKEVMIGTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service