Skip to Content
Merck
All Photos(1)

Key Documents

D205206

Sigma-Aldrich

N,N′-Diphenylbenzidine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€97.70

€97.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€97.70

About This Item

Linear Formula:
4-(C6H5NH)C6H4C6H4-4-(NHC6H5)
CAS Number:
Molecular Weight:
336.43
Beilstein:
2220993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
97%

€97.70


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

technique(s)

titration: suitable

mp

246-248 °C (lit.)

SMILES string

N(c1ccccc1)c2ccc(cc2)-c3ccc(Nc4ccccc4)cc3

InChI

1S/C24H20N2/c1-3-7-21(8-4-1)25-23-15-11-19(12-16-23)20-13-17-24(18-14-20)26-22-9-5-2-6-10-22/h1-18,25-26H

InChI key

FDRNXKXKFNHNCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′-Diphenylbenzidine can be employed as a building block for in the preparation of hole-transporting materials for organic LEDs.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characteristics of Poly [N, N′ -diphenyl-N, N′ -bis (4-aminobiphenyl)-(1, 1′ -biphenyl)-4, 4′ -diamine pyromellitimide] as a Hole Injecting and Transporting Layer for Hybrid Organic Light-Emitting Device
Kim Youngkyoo, et al.
Macromolecules, 35(23), 8759-8767 (2002)
Amorphous molecular materials: charge transport in the glassy state of N, N′ -di (biphenylyl)-N, N′ -diphenyl-[1, 1′ -biphenyl]-4, 4′ -diamines
Okumoto K, et al.
Synthetic Metals, 111(23), 473-476 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service