Skip to Content
Merck
All Photos(2)

Key Documents

D142158

Sigma-Aldrich

Dimethyl 5-aminoisophthalate

98%

Synonym(s):

Dimethyl 5-aminobenzene-1,3-dicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€152.00

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€152.00

About This Item

Linear Formula:
H2NC6H3-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
209.20
Beilstein:
2116565
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

178-181 °C (lit.)

SMILES string

COC(=O)c1cc(N)cc(c1)C(=O)OC

InChI

1S/C10H11NO4/c1-14-9(12)6-3-7(10(13)15-2)5-8(11)4-6/h3-5H,11H2,1-2H3

InChI key

DEKPYXUDJRABNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuejuan Ma et al.
Journal of hazardous materials, 355, 65-73 (2018-05-19)
The degradation of organophosphorous nerve agents is of primary concern due to the severe toxicity of these agents. Based on the active center of organophosphorus hydrolase (OPH), a bimetallic nuclear ligand, (5-vinyl-1,3-phenylene)bis(di(1H-imidazol-2-yl) methanol) (VPIM), was designed and synthesized, which contains
Vijay Gupta et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(12), 2658-2665 (2019-12-04)
A new triazole-functionalized tetracarboxylic acid ligand (H4 L) has been synthesized and utilized for the fabrication of a 3D ZnII organic framework with a Zn4 (-COO)6 cluster as the secondary building unit. The framework exhibits very good thermal stability and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service