Key Documents
CLS999818
Corning® phenolic cap, PTFE liner
18-415 thread
Synonym(s):
black phenolic, phenolic caps, screw caps, Corning® 9998
Select a Size
192 EA
€134.00
€134.00
Please contact Customer Service for Availability
Select a Size
Change View
192 EA
€134.00
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
€134.00
Please contact Customer Service for Availability
material
PTFE liner
black phenolic cap
manufacturer/tradename
Corning 9998-18
H
~17.5 mm
fitting
18-415 thread
General description
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)