CLS948036
Pyrex® Filter tube/thimble
I.D. × L 36 mm × 160 mm
Synonym(s):
Corning® 9480
About This Item
Recommended Products
material
borosilicate glass
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 9480-36
I.D. × L
36 mm × 160 mm
filter diam.
36 mm
stem O.D. × L
8 mm × 75 mm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)