CLS7078D1
Pyrex® serological pipettes, disposable, flip-top canister pack, sterile
volume 1.0 mL
Synonym(s):
pipet
Select a Size
€285.00
Select a Size
About This Item
€285.00
Recommended Products
material
borosilicate glass
sterility
sterile
feature
subdivision 0.01 mL
manufacturer/tradename
Corning 7078D-1
volume measuring range
1.0 mL
capacity
1 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service