Skip to Content
Merck
All Photos(1)

Documents

CLS694411L

Pyrex® rectangular chromatography jar

L × W × H 181 mm × 238 mm × 324 mm

Synonym(s):

Corning® 6944

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

grade

for chromatography

manufacturer/tradename

Corning 6944-11L

L × W × H

181 mm × 238 mm × 324 mm

capacity

11.4 L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ground to the close tolerances needed for tight cover fit. Jar edges are ground flat within 0.25mm. Covers are not supplied. Do not use heat, pressure or vacuum applications.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service