CLS56421L
Pyrex® volumetric flasks with PE stopper
capacity 1,000 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5642
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
long
stopper size
22
sterility
non-sterile
feature
class A
closure type Polyethylene stopper
packaging
pack of 1
case of 6
manufacturer/tradename
Corning 5642-1L
tol. measuring range
0.30 mL
capacity
1,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® volumetric flasks feature machine-blown bodies with sealed heavy-beaded, heavy-tubing necks.
- The linear high-density polyethylene stoppers are made with a closed bottom to conform to Standard Taper stopper dimensions.
- A sharp permanent graduation line and large white block letters facilitate accurate volumetric measurements.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service