CLS5641200
Pyrex® volumetric flask, with Pyrex® ST stopper
capacity 200 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5641
About This Item
Recommended Products
stopper size
13
feature
class B
packaging
case of 12 ea
case of 6 ea
manufacturer/tradename
Corning 5641-200
tol. measuring range
0.20 mL
capacity
200 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service