Key Documents
CLS4450250
Pyrex® baffled Erlenmeyer flask
capacity 250 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, laboratory flasks, Corning® 4450
About This Item
UNSPSC Code:
41122104
eCl@ss:
32040501
Product CLS4450250 is not currently sold in your country. Contact Technical Service
material
borosilicate glass
flat bottom flask
narrow-mouth
narrow-mouth neck
sterility
non-sterile
feature
cap: no
closure type Rubber stopper
packaging
case of 6
manufacturer/tradename
Corning 4450-250
capacity
250 mL
General description
  • Pyrex® baffled Erlenmeyer flasks feature a heavy tooled rim for maximum durability.
  • Baffle-designed Delong flasks promote optimal and consistent solution agitation when used with flask shakers.
  • Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.
  • Includes a marking spot for convenience.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)