Skip to Content
Merck
All Photos(1)

Key Documents

CLS139510L

Pyrex® round media storage bottles and reusable screw caps

capacity 10,000 mL

Synonym(s):

Pyrex® media bottles, bottle, glass media bottles, laboratory bottles, media bottle, pyrex bottle, round bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
Product CLS139510L is not currently sold in your country. Contact Technical Service

material

borosilicate glass bottle
clear bottle
cylindrical bottle
orange polypropylene cap (plug seal)
wide-mouth bottle

sterility

non-sterile

feature

autoclavable
cap
graduations
reusable

packaging

case of 10

manufacturer/tradename

Corning 1395-10L

availability

not available in EU

capacity

10,000 mL

diam. × H

234 mm × 410 mm

fitting

GL 45 neck thread

Looking for similar products? Visit Product Comparison Guide

General description

Pyrex heavy-duty round media storage bottles can be used for storage as well as for mixing and sampling. These media storage bottles Include linerless, one-piece, autoclavable, orange PP plug-seal caps with drip-free pouring rings. Optional red PBTP caps and red ETFE pouring rings are available separately, for dry heat sterilization (180°C).

Pyrex round media storage bottles are made of borosilicate glass with permanent, white enamel graduations and marking spots.

Caution: Bottles larger than 2 L should NOT be used with bottle-top filter units or in other applications involving vacuum/pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

A wide range of optional caps are available for these bottles:
  • Vented membrane cap with 0.22 μm PTFE hydrophobic membrane
  • Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C)
  • PBT caps with silicone septa for syringe sampling or introduction of reagents
These heavy duty 10 L PYREX round bottles can be used for storage as well as mixing and sampling. They include a linerless, one-piece autoclavable orange polypropylene plug seal GL45 threaded cap with a drip-free pouring ring. Bottles have permanent white enamel graduations and marking spots.

Caution: Bottles larger than 2 L should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. A vented cap is NOT recommended for use on the 10 L 1395 bottle for venting during autoclaving. -A wide range of optional caps are available for these bottles: -Polypropylene colored caps in a choice of orange, green, purple, red, and light gray for ease of sorting and identification. -Vented membrane cap with 0.22 μm PTFE hydrophobic membrane. -Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C). -PBT caps with silicone septa for syringe sampling or introduction of reagents.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service