Skip to Content
Merck
All Photos(1)

Key Documents

CF0001

Sigma-Aldrich

1-Chloro-3,3-dimethylbenziodoxole

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€220.00

€220.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€220.00

About This Item

Linear Formula:
C9H10ClIO
CAS Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€220.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

[I]1(OC(c2c1cccc2)(C)C)Cl

InChI

1S/C9H10ClIO/c1-9(2)7-5-3-4-6-8(7)11(10)12-9/h3-6H,1-2H3

InChI key

HEBYOYIIQNKPGQ-UHFFFAOYSA-N

Application

1-Chloro-3,3-dimethylbenziodoxole is a crystalline, air-stable, yellow chloroiodane compound which finds use as a starting material for production of other hypervalent iodine compounds. It can also be used as a mild chlorinating reagent. The compound shows good solubility in polar organic solvents, typically dichloromethane or ethyl acetate.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service