Skip to Content
Merck
All Photos(1)

Documents

CDS009203

Sigma-Aldrich

3-o-Ethyl-L-ascorbic acid

AldrichCPR

Synonym(s):

3-O-Ethyl-L-ascorbic acid, 3-O-Ethylascorbic acid, L-Ascorbic acid 3-O-ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O6
CAS Number:
Molecular Weight:
204.18
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:

form

solid

SMILES string

CCOC1=C(O)C(=O)O[C@@H]1[C@@H](O)CO

InChI

1S/C8H12O6/c1-2-13-7-5(11)8(12)14-6(7)4(10)3-9/h4,6,9-11H,2-3H2,1H3/t4-,6+/m0/s1

InChI key

ZGSCRDSBTNQPMS-UJURSFKZSA-N

Application

  • Whitening Cream Formulation: 3-O-Ethyl-L-ascorbic acid is used in whitening cream products, highlighting its importance in ensuring product safety and efficacy (Permana et al., 2023).
  • Oral Curcumin Delivery: 3-O-Ethyl-L-Ascorbic Acid is used to doped enteric-coated gelatin capsules that aimed at enhancing oral curcumin delivery for cancer treatment, demonstrating its potential in biomedical applications (Lim, 2022).
  • Dental Enamel Treatment: Used in a one-step surface treatment reagent with 50% citric acid solution for restoring the shear bond strength of metal brackets bonded to bleached human enamel, proposing its use in orthodontic treatments (Saeoweiang et al., 2023).
  • Advanced Skin Pigmentation Therapy: 3-O-Ethyl-L-ascorbic acid is used for treating skin pigmentation disorders, suggesting a new therapeutic approach in dermatology (Yan et al., 2023).
  • Cell Viability and Melanin Synthesis: A study explored the multifaceted effects of 3-O-Ethyl-L-ascorbic acid, on the viability and melanin synthesis of B16/F10 cells under various conditions, indicating its role in melanogenesis and potential cosmetic applications (Choi et al., 2024).

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICHā€²S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service