Skip to Content
Merck
All Photos(1)

Key Documents

CDS000584

Sigma-Aldrich

Betanin

AldrichCPR

Synonym(s):

Red beet extract diluted with Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O13
Molecular Weight:
550.47
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O[C@H]1[C@H](O)[C@@H](O)[C@H](OC2=CC(CC(C([O-])=O)/[N+]3=C\C=C4C=C(C(O)=O)NC(C(O)=O)C\4)=C3C=C2O)O[C@@H]1CO

InChI

1S/C24H26N2O13/c27-8-17-18(29)19(30)20(31)24(39-17)38-16-6-10-5-14(23(36)37)26(13(10)7-15(16)28)2-1-9-3-11(21(32)33)25-12(4-9)22(34)35/h1-3,6-7,12,14,17-20,24,27,29-31H,4-5,8H2,(H4,28,32,33,34,35,36,37)/t12?,14?,17-,18-,19+,20-,24-/m1/s1

InChI key

DHHFDKNIEVKVKS-UWTTYFQQSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elham Hadipour et al.
Phytotherapy research : PTR, 34(8), 1847-1867 (2020-03-15)
Over the past few years, the use of natural substances as protective or therapeutic agents has gained much attention worldwide. Recent modern studies have shown a variety of health benefits for red beetroot and its active compounds betalains (also betanin)
Charng-Cherng Chyau et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
The aim of this study was to provide new insights into the role of the ethanolic extracts of Djulis (Chenopodium formosanum, EECF) and its bioactive compounds in preventing adipogenesis in 3T3-L1 adipocytes. The results demonstrated EECF significantly inhibited oil red
Haizhao Song et al.
Journal of agricultural and food chemistry, 64(1), 236-244 (2015-12-15)
Natural bioactive compounds in food have been shown to be beneficial in preventing the development of obesity, diabetes, and other metabolic diseases. Increasing evidence indicates that betacyanins possess free-radical-scavenging and antioxidant activities, suggesting their beneficial effects on metabolic disorders. The
Kah Yee Choo et al.
Food science and biotechnology, 27(5), 1411-1417 (2018-10-16)
The objective of this work was to study the effect of storage temperatures and duration on the stability of fermented red dragon fruit drink (FRDFD) on its betacyanins content, physicochemical and microbiological qualities (BPM) and determining sensory acceptability. Results showed
Marco Tutone et al.
Journal of theoretical biology, 455, 147-160 (2018-07-22)
Indicaxanthin is a bioactive and bioavailable betalain pigment extracted from Opuntia ficus indica fruits. Indicaxanthin has pharmacokinetic proprieties, rarely found in other phytochemicals, and it has been demonstrated that it provides a broad-spectrum of pharmaceutical activity, exerting anti-proliferative, anti-inflammatory, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service