Skip to Content
Merck
All Photos(1)

Key Documents

C98403

Sigma-Aldrich

Cycloheptane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14
CAS Number:
Molecular Weight:
98.19
Beilstein:
1900279
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

vapor pressure

44 mmHg ( 37.7 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

118.5 °C (lit.)

mp

−12 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

SMILES string

C1CCCCCC1

InChI

1S/C7H14/c1-2-4-6-7-5-3-1/h1-7H2

InChI key

DMEGYFMYUHOHGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jossue Ortiz-Álvarez et al.
Journal of basic microbiology, 59(8), 792-806 (2019-08-02)
The aim of this study was to examine four strains of two yeast species in relation to their capability for assimilating alkanes in the presence of heavy metals (HMs). The four strains tested were Candida pseudoglaebosa ENCB-7 and Kodamaea ohmeri
Chris E Freye et al.
Talanta, 211, 120668-120668 (2020-02-20)
The ability to discover minute differences between samples or sample classes for gas chromatography coupled to mass spectrometry (GC-MS) can be a challenging endeavor, especially when those differences are not a priori. Fisher ratio (F-ratio) analysis is an apt technique
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
Michael H Abraham et al.
European journal of medicinal chemistry, 44(2), 885-890 (2008-07-08)
The convulsant activity of 48 compounds studied by Eger et al. has been analyzed using an Abraham solvation equation. Four compounds identified by Eger et al. as more potent than expected were similarly identified, and for the remaining 44 compounds
A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service