Skip to Content
Merck
All Photos(1)

Documents

C97482

Sigma-Aldrich

trans,trans,cis-1,5,9-Cyclododecatriene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18
CAS Number:
Molecular Weight:
162.27
Beilstein:
1362442
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.507 (lit.)

bp

231 °C (lit.)

mp

−18 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

C1C\C=C\CC\C=C\CCC=C1

InChI

1S/C12H18/c1-2-4-6-8-10-12-11-9-7-5-3-1/h1-2,7-10H,3-6,11-12H2/b2-1-,9-7+,10-8+

InChI key

ZOLLIQAKMYWTBR-RYMQXAEESA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reinout Van Kerrebroeck et al.
Molecules (Basel, Switzerland), 24(11) (2019-06-07)
Bromination reactions are crucial in today's chemical industry since the versatility of the formed organobromides makes them suitable building blocks for numerous syntheses. However, the use of the toxic and highly reactive molecular bromine (Br2) makes these brominations very challenging
Xianmiao Zhang et al.
Environmental science. Processes & impacts, 22(4), 1026-1036 (2020-03-03)
The reactions of hexabromocyclododecane (HBCD) isomers with Fe(ii) associated with iron oxides were performed in a pH range from 6.15 to 7.50 at room temperature. It was observed that Fe(ii) associated with iron oxides (i.e., goethite, magnetite, hematite) is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service