Skip to Content
Merck
All Photos(1)

Key Documents

C76604

Sigma-Aldrich

Cholesteryl chloride

97%

Synonym(s):

3β-Chloro-5-cholestene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45Cl
CAS Number:
Molecular Weight:
405.10
Beilstein:
2703655
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid crystal

optical activity

[α]25/D −24°, c = 1 in chloroform

mp

94-96 °C (lit.)

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](Cl)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H45Cl/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

OTVRYZXVVMZHHW-DPAQBDIFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Weber et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 32(4), 297-303 (1994-04-01)
3 beta-Chlorosteroids, such as cholesteryl beta-chloride and sitosteryl beta-chloride, are formed during the production of protein hydrolysates, which are useful flavour enhancers. These chlorinated steroids may also attract attention as environmental contaminants if they are released from liquid crystal display
P Oswald
The European physical journal. E, Soft matter, 35(2), 10-10 (2012-02-22)
In a recent letter (EPL 97, 36006 (2012)), we have shown that the Leslie thermomechanical coupling cannot alone explain the Lehmann effect (namely the rotation of cholesteric droplets when they are subjected to a temperature gradient). This result was obtained
N Weber
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 27(4), 259-263 (1989-04-01)
A method for the analysis of 3 beta-chloro steroids by high-performance liquid chromatography is described. These compounds are known to occur in commercial protein hydrolysates. The gastro-intestinal absorption, distribution and metabolism of chlorinated steroids were studied after their intragastric application
Kamlesh Sharma et al.
Journal of molecular modeling, 11(2), 135-140 (2005-04-20)
This work describes a theoretical approach to the substitution reaction mechanism involving the conversion of cholesterol to cholesteryl chloride. Two chlorosulfite ester molecules were formed as intermediates. An iso-steroid was found as the transition state. The final product was cholesteryl
Smiljka S Milisavljević et al.
Steroids, 70(13), 867-872 (2005-07-26)
Four 5,6-unsaturated steroids--3beta-chlorocholest-5-ene (1a), cholesterol (1b) and its acetate (1c) and benzoate (1d)-were subjected to constant current electrolysis (50 mA, 2 F mol(-1)) in an electrolytic cell divided by a ceramic membrane, using a platinum foil as the anode and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service