Skip to Content
Merck
All Photos(1)

Key Documents

C106003

Sigma-Aldrich

(Bromomethyl)cyclohexane

99%

Synonym(s):

Cyclohexylmethyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€38.80
25 G
€51.10
100 G
€118.00

€38.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€38.80
25 G
€51.10
100 G
€118.00

About This Item

Linear Formula:
C6H11CH2Br
CAS Number:
Molecular Weight:
177.08
Beilstein:
635738
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€38.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

76-77 °C/26 mmHg (lit.)

density

1.269 g/mL at 25 °C (lit.)

SMILES string

BrCC1CCCCC1

InChI

1S/C7H13Br/c8-6-7-4-2-1-3-5-7/h7H,1-6H2

InChI key

UUWSLBWDFJMSFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Doi et al.
Forensic toxicology, 36(1), 51-60 (2018-01-26)
Recently, carboxamide-type synthetic cannabinoids have been distributed globally as new psychoactive substances (NPS). Some of these compounds possess asymmetric carbon, which is derived from an amide moiety composed of amino acid derivatives (i.e., amides or esters of amino acids). In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service