BR781665
BRAND® microplate BRANDplates®, pureGrade
size 96 wells, size 350 μL, flat bottom, white
Synonym(s):
BRANDplates®, Microplates, PCR, Plates, Sealing films, pureGradeS
Select a Size
€240.00
Select a Size
About This Item
polystyrene
white wells
96 wells
€240.00
Recommended Products
material
flat bottom
polystyrene
white wells
sterility
sterile
feature
lid
skirt (F-bottom)
packaging
pack of 50 ea
pkg of (individually packaged)
manufacturer/tradename
BRAND 781665
size
350 μL
96 wells
working volume
350 μL
color
white
suitability
suitable for (bacteriological assays)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- BRAND® microplate BRANDplates® pureGrade are white flat bottom standard type plates of BIO-CERT® CERTIFIED QUALITY.
- Optimal surfaces and Reliable quality.
- Non-treated surface and β-radiation sterilized.
- Applicable for bacteriological assays.
- Non-cytotoxic and free from DNase, DNA, RNase and endotoxins.
Features and Benefits
- Optimal surfaces for optimal results
- Reliable quality from the cleanroom
- Tested by independent laboratories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service