BR747730
BRAND® Pasteur Pipette
soda-lime glass, capacity × L ~1.5 mL × 310 mm, Un-plugged
Select a Size
€181.00
Select a Size
About This Item
€181.00
Recommended Products
material
soda-lime glass
sterility
non-sterile
feature
graduation: no
Un-plugged
packaging
pack of 2 ea
manufacturer/tradename
BRAND 747730
capacity × L
~1.5 mL × 310 mm
volume
~1.5 mL , Withdraw
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service